Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001175.6 | NP_001166.3 | 2 | 305 | |
NM_001321420.1 | NP_001308349.1 | 3 | 463 | ||
NM_001321421.1 | NP_001308350.1 | 2 | 184 | ||
NM_001321422.1 | NP_001308351.1 | 2 | 306 | ||
NM_001321423.1 | NP_001308352.1 | 2 | 172 | ||
GenBank mRNA | AA143425.1 | - | 2 | 113 | |
AB451315.1 | - | 1 | 59 | ||
AB451445.1 | - | 1 | 59 | ||
AF498927.1 | - | 1 | 59 | ||
AK311807.1 | - | 2 | 135 | ||
AV685495.1 | - | 2 | 164 | ||
BC009200.2 | - | 2 | 109 | 3621138 | |
BE744996.1 | - | 2 | 170 | ||
BG390701.1 | - | 3 | 280 | ||
BI762648.1 | - | 2 | 304 | ||
BP355037.1 | - | 2 | 280 | ||
BY996333.1 | - | 2 | 169 | ||
CD385175.1 | - | 2 | 323 | ||
L07916.1 | - | 2 | 133 | ||
L20688.1 | - | 2 | 211 | ||
X69549.1 | - | 2 | 113 |