Gene Transcripts

Gene Symbol
C5
Entrez Gene ID
727
Gene Name
complement component 5
Gene Aliases
C5D, C5a, C5b, CPAMD4, ECLZB
Chromosome Location
Chr. 9: 120952335 - 121075174 on Build GRCh38
UniGene ID
Hs.494997
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317163.1 NP_001304092.1 19 2724
NM_001317164.1 NP_001304093.1 19 2395
NM_001735.2 NP_001726.2 19 2398
XM_011518980.2 XP_011517282.1 20 2767
XM_017015102.1 XP_016870591.1 21 2919
XM_017015103.1 XP_016870592.1 19 2552
XM_017015104.1 XP_016870593.1 21 2919
GenBank mRNA AB209031.1 - 19 2724
AK310774.1 - 19 2395
BC113738.1 - 19 2393 8322790
BC113740.1 - 19 2393 8322792
M57729.1 - 19 2380
M65134.1 - 9 1135
T82068.1 - 2 114

View Full Product Details