Gene Transcripts

Gene Symbol
OR3A2
Entrez Gene ID
4995
Gene Name
olfactory receptor family 3 subfamily A member 2
Gene Aliases
OLFRA04, OR17-14, OR17-228, OR228
Chromosome Location
Chr. 17: 3276113 - 3386311 on Build GRCh38
UniGene ID
Hs.647414
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002551.3 NP_002542.3 1 833
XM_017024697.1 XP_016880186.1 6 1453
XM_017024698.1 XP_016880187.1 4 1145
GenBank mRNA BC095517.1 - 1 833

View Full Product Details