Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_032242.3 | NP_115618.3 | 22 | 4310 | |
XM_011512908.2 | XP_011511210.1 | 23 | 4412 | ||
XM_011512909.1 | XP_011511211.1 | 23 | 5005 | ||
GenBank mRNA | AK293425.1 | - | 3 | 429 | |
X87832.2 | - | 22 | 4245 |