Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001076785.2 | NP_001070253.1 | 10 | 1528 | |
NM_003983.5 | NP_003974.3 | 9 | 1441 | ||
XM_011523433.1 | XP_011521735.1 | 9 | 1543 | ||
XM_011523434.1 | XP_011521736.1 | 9 | 1608 | ||
XM_011523438.1 | XP_011521740.1 | 8 | 1998 | ||
XM_011523439.1 | XP_011521741.1 | 10 | 2240 | ||
XM_017023851.1 | XP_016879340.1 | 8 | 1568 | ||
GenBank mRNA | AK310602.1 | - | 5 | 506 | |
AK311610.1 | - | 5 | 802 | ||
BC028216.1 | - | 9 | 1397 | 5214340 | |
CR749291.1 | - | 9 | 1427 | ||
CR749475.1 | - | 8 | 1265 | ||
D87432.1 | - | 9 | 1476 |