Gene Transcripts

Gene Symbol
ACAP1
Entrez Gene ID
9744
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 1
Gene Aliases
CENTB1
Chromosome Location
Chr. 17: 7336506 - 7354944 on Build GRCh38
UniGene ID
Hs.337242
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014716.3 NP_055531.1 20, 21 2282
GenBank mRNA BC018543.1 - 20, 21 2257
BT009788.1 - 20, 21 2076
D30758.1 - 20, 21 2264

View Full Product Details