Gene Transcripts

Gene Symbol
SH2B3
Entrez Gene ID
10019
Gene Name
SH2B adaptor protein 3
Gene Aliases
IDDM20, LNK
Chromosome Location
Chr. 12: 111405106 - 111451623 on Build GRCh38
UniGene ID
Hs.506784
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291424.1 NP_001278353.1 4 440
NM_005475.2 NP_005466.1 5 1363
XM_005253818.4 XP_005253875.1 6 1507
XM_005253819.4 XP_005253876.1 5 1390
XM_006719180.3 XP_006719243.1 4 992
XM_011537719.2 XP_011536021.1 6 1510
XM_011537720.2 XP_011536022.1 6 1175
XM_011537721.2 XP_011536023.1 4 2975
GenBank mRNA AB208911.1 - 4 599
AF055581.1 - 5 1363
AJ012793.1 - 4 1009
AK302173.1 - 4 440
BC136451.1 - 5 1049 9020440

View Full Product Details