Gene Transcripts

Gene Symbol
ARPC4
Entrez Gene ID
10093
Gene Name
actin related protein 2/3 complex subunit 4
Gene Aliases
ARC20, P20-ARC
Chromosome Location
Chr. 3: 9792495 - 9807105 on Build GRCh38
UniGene ID
Hs.323342
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024959.2 NP_001020130.1 2 298
NM_001024960.2 NP_001020131.1 2 153
NM_001198780.1 NP_001185709.1 2 152
NM_005718.4 NP_005709.1 2 664
GenBank mRNA AF006087.1 - 2 105
AF019888.1 - 1 90
AW408422.1 - 2 111
BC012596.2 - 1 87
BG938423.1 - 2 136
BI768875.1 - 2 277
BI910730.1 - 2 672
BM045872.1 - 2 163
BQ688329.1 - 2 236
BX419672.2 - 2 152
DA226584.1 - 2 153
DB060690.1 - 2 298

Gene Symbol
ARPC4-TTLL3
Entrez Gene ID
100526693
Gene Name
ARPC4-TTLL3 readthrough
Gene Aliases
Chromosome Location
Chr. 3: 9792548 - 9836356 on Build GRCh38
UniGene ID
Hs.323342
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001198793.1 NP_001185722.1 2 664
GenBank mRNA AK024110.1 - 2 654
BC025289.1 - 1 98

View Full Product Details