Gene Transcripts

Gene Symbol
SSSCA1
Entrez Gene ID
10534
Gene Name
Sjogren syndrome/scleroderma autoantigen 1
Gene Aliases
p27
Chromosome Location
Chr. 11: 65570459 - 65571888 on Build GRCh38
UniGene ID
Hs.25723
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006396.2 NP_006387.1 1, 2 51
GenBank mRNA AB001740.1 - 1, 2 38
BC014791.2 - 1, 2 37 3857068
BP351453.1 - 1, 2 52

View Full Product Details