Gene Transcripts
- Gene Symbol
- CD226
- Entrez Gene ID
- 10666
- Gene Name
- CD226 molecule
- Gene Aliases
- DNAM-1, DNAM1, PTA1, TLiSA1
- Chromosome Location
- Chr. 18: 69860228 - 69962086 on Build GRCh38
- UniGene ID
- Hs.660130
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001303618.1 |
NP_001290547.1
|
3, 4 |
1165 |
|
NM_001303619.1 |
NP_001290548.1
|
2, 3 |
758 |
|
NM_006566.3 |
NP_006557.2
|
4, 5 |
1167 |
|
XM_005266642.3 |
XP_005266699.1
|
4, 5 |
1266 |
|
XM_005266643.3 |
XP_005266700.1
|
2, 3 |
360 |
|
XM_006722374.3 |
XP_006722437.1
|
2, 3 |
831 |
|
XM_017025524.1 |
XP_016881013.1
|
4, 5 |
7910 |
|
XM_017025525.1 |
XP_016881014.1
|
2, 3 |
642 |
|
XM_017025526.1 |
XP_016881015.1
|
3, 4 |
536 |
|
XM_017025527.1 |
XP_016881016.1
|
2, 3 |
1200 |
|
GenBank mRNA |
AJ537553.1 |
- |
3, 4 |
747 |
|
AK313199.1 |
- |
3, 4 |
1166 |
|
BC074787.2 |
- |
3, 4 |
752 |
30915562 |
GU935335.1 |
- |
4, 5 |
914 |
|
GU935336.1 |
- |
2, 3 |
517 |
|
U56102.1 |
- |
4, 5 |
926 |
|
View Full Product Details