Gene Transcripts

Gene Symbol
SPACA9
Entrez Gene ID
11092
Gene Name
sperm acrosome associated 9
Gene Aliases
C9orf9, Mast
Chromosome Location
Chr. 9: 132878320 - 132890314 on Build GRCh38
UniGene ID
Hs.62595
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001316897.1 NP_001303826.1 3 476
NM_001316898.1 NP_001303827.1 3 469
NM_001316900.1 NP_001303829.1 3 476
NM_018956.4 NP_061829.3 3 476
XM_017014231.1 XP_016869720.1 3 472
GenBank mRNA AJ011375.1 - 3 464
AK075259.1 - 3 469
BC012940.1 - 3 426 4478288
CK906174.1 - 3 251
CK906175.1 - 3 422
CR457210.1 - 2 315
DA134573.1 - 3 469

View Full Product Details