Gene Transcripts

Gene Symbol
ARSG
Entrez Gene ID
22901
Gene Name
arylsulfatase G
Gene Aliases
Chromosome Location
Chr. 17: 68259089 - 68452000 on Build GRCh38
UniGene ID
Hs.437249
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267727.1 NP_001254656.1 9 1678
NM_014960.4 NP_055775.2 9 1849
XM_005257170.3 XP_005257227.1 9 1862
XM_006721777.3 XP_006721840.2 9 1876
XM_011524535.2 XP_011522837.1 9 1874
XM_011524536.2 XP_011522838.1 9 1704
XM_011524537.1 XP_011522839.1 9 1625
XM_011524538.2 XP_011522840.1 9 1876
XM_011524540.2 XP_011522842.1 9 1896
XM_011524541.2 XP_011522843.1 9 1942
XM_011524542.2 XP_011522844.1 9 1942
XM_011524543.2 XP_011522845.1 9 1942
XM_011524544.2 XP_011522846.1 9 1942
XM_011524545.2 XP_011522847.1 9 1900
XM_011524546.2 XP_011522848.1 4 547
XM_017024360.1 XP_016879849.1 9 1714
XM_017024361.1 XP_016879850.1 9 1862
XM_017024362.1 XP_016879851.1 9 1801
XM_017024363.1 XP_016879852.1 9 1707
XM_017024364.1 XP_016879853.1 9 1703
XM_017024365.1 XP_016879854.1 9 1691
XM_017024366.1 XP_016879855.1 9 1902
XM_017024367.1 XP_016879856.1 9 1704
XM_017024368.1 XP_016879857.1 9 1704
GenBank mRNA AB023218.1 - 8 1511
AY358380.1 - 9 1849
BC012375.1 - 8 1092 3882163

View Full Product Details