Gene Transcripts

Gene Symbol
ZCCHC14
Entrez Gene ID
23174
Gene Name
zinc finger CCHC-type containing 14
Gene Aliases
BDG-29, BDG29
Chromosome Location
Chr. 16: 87406246 - 87492948 on Build GRCh38
UniGene ID
Hs.156231
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015144.2 NP_055959.1 2 218
XM_005255858.3 XP_005255915.2 2 409
XM_011522964.2 XP_011521266.1 2 1312
XM_017023082.1 XP_016878571.1 2 1168
GenBank mRNA AB011151.2 - 2 76
AB030243.1 - 2 226
AK289682.1 - 2 409
BC101478.1 - 2 225 8068984

View Full Product Details