Gene Transcripts

Gene Symbol
MLC1
Entrez Gene ID
23209
Gene Name
megalencephalic leukoencephalopathy with subcortical cysts 1
Gene Aliases
LVM, MLC, VL
Chromosome Location
Chr. 22: 50059391 - 50085929 on Build GRCh38
UniGene ID
Hs.517729
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015166.3 NP_055981.1 8, 9 1344
NM_139202.2 NP_631941.1 8, 9 983
XM_011530678.1 XP_011528980.1 8, 9 1105
XM_017028671.1 XP_016884160.1 7, 8 772
GenBank mRNA AF319633.1 - 8, 9 906
AK124264.1 - 8, 9 1345
AK294048.1 - 6, 7 734
AK295106.1 - 7, 8 605
AK297341.1 - 7, 8 704
AK299841.1 - 7, 8 752
BC028425.2 - 8, 9 943 4817175
BC070042.1 - 8, 9 941 5285102
CR456460.1 - 7, 8 762
D25217.3 - 8, 9 1020

View Full Product Details