Gene Transcripts

Gene Symbol
RUSC1
Entrez Gene ID
23623
Gene Name
RUN and SH3 domain containing 1
Gene Aliases
NESCA
Chromosome Location
Chr. 1: 155320849 - 155331118 on Build GRCh38
UniGene ID
Hs.226499
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105203.1 NP_001098673.1 8 2579
NM_001105204.1 NP_001098674.1 8 2261
NM_001105205.1 NP_001098675.1 7 1601
NM_001278227.1 NP_001265156.1 7 1545
NM_001278228.1 NP_001265157.1 6 1781
NM_001278229.1 NP_001265158.1 7 1326
NM_001278230.1 NP_001265159.1 6 1443
NM_014328.4 NP_055143.2 7 1644
NR_103478.1 6 991
XM_006711254.1 XP_006711317.1 9 2990
XM_006711256.1 XP_006711319.1 9 2672
XM_006711257.1 XP_006711320.1 7 1258
XM_017000891.1 XP_016856380.1 8 3160
XM_017000892.1 XP_016856381.1 7 940
GenBank mRNA AB026894.2 - 7 1068
AK055451.1 - 7 1839
AK074485.1 - 6 879
AK074982.1 - 7 1063
AK093777.1 - 5 1903
AK125378.1 - 8 2213
AK298730.1 - 7 1055
AK307736.1 - 6 1230
AK314559.1 - 7 1313
BC001045.2 - 7 1085
BC004293.1 - 6 847
BC028055.1 - 6 1193
BC052277.1 - 7 1601
BX402179.2 - 7 817
BX640612.1 - 6 1261
CR749671.1 - 6 2150

View Full Product Details