Gene Transcripts

Gene Symbol
CNPPD1
Entrez Gene ID
27013
Gene Name
cyclin Pas1/PHO80 domain containing 1
Gene Aliases
C2orf24, CGI-57
Chromosome Location
Chr. 2: 219171897 - 219178174 on Build GRCh38
UniGene ID
Hs.4973
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321389.1 NP_001308318.1 5 721
NM_001321390.1 NP_001308319.1 5 366
NM_001321391.1 NP_001308320.1 5 400
NM_015680.5 NP_056495.3 4 458
NR_135628.1 5 400
NR_135629.1 4 458
GenBank mRNA AF151815.1 - 5 625
AK075274.1 - 4 414
AK314975.1 - 4 401
AY007160.1 - 4 431
BC001393.2 - 4 381 3051397
BC012821.2 - 4 430 2958621
BI912613.1 - 5 367
CR457401.1 - 4 293
CX166359.1 - 5 717
DB477110.1 - 4 445

View Full Product Details