Gene Transcripts
- Gene Symbol
- NME7
- Entrez Gene ID
- 29922
- Gene Name
- NME/NM23 family member 7
- Gene Aliases
- CFAP67, MN23H7, NDK 7, NDK7, nm23-H7
- Chromosome Location
- Chr. 1: 169132385 - 169367963 on Build GRCh38
- UniGene ID
- Hs.706952
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013330.4 |
NP_037462.1
|
7, 8 |
1000 |
|
NM_197972.2 |
NP_932076.1
|
7, 8 |
969 |
|
NR_104229.1 |
|
7, 8 |
1000 |
|
XM_005245106.4 |
XP_005245163.1
|
5, 6 |
578 |
|
XM_017001099.1 |
XP_016856588.1
|
7, 8 |
986 |
|
XM_017001100.1 |
XP_016856589.1
|
5, 6 |
566 |
|
GenBank mRNA |
AF153191.2 |
- |
7, 8 |
839 |
|
AK094513.1 |
- |
7, 8 |
952 |
|
AK290701.1 |
- |
7, 8 |
834 |
|
AK301917.1 |
- |
7, 8 |
983 |
|
BC006983.1 |
- |
7, 8 |
831 |
3997152 |
View Full Product Details