Gene Transcripts
- Gene Symbol
- ANAPC4
- Entrez Gene ID
- 29945
- Gene Name
- anaphase promoting complex subunit 4
- Gene Aliases
- APC4
- Chromosome Location
- Chr. 4: 25377226 - 25418498 on Build GRCh38
- UniGene ID
- Hs.152173
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286756.1 |
NP_001273685.1
|
9, 10 |
842 |
|
NM_013367.2 |
NP_037499.2
|
9, 10 |
842 |
|
XM_011513838.1 |
XP_011512140.1
|
9, 10 |
719 |
|
XM_011513839.1 |
XP_011512141.1
|
9, 10 |
763 |
|
XM_017008138.1 |
XP_016863627.1
|
9, 10 |
763 |
|
GenBank mRNA |
AF191338.1 |
- |
8, 9 |
713 |
|
AK023597.1 |
- |
6, 7 |
453 |
|
AK292336.1 |
- |
9, 10 |
762 |
|
AY429517.1 |
- |
2, 3 |
190 |
|
AY429518.1 |
- |
2, 3 |
177 |
|
BC059383.1 |
- |
9, 10 |
815 |
30349524 |
View Full Product Details