Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199583.2 | NP_001186512.1 | 8 | 1211 | |
NM_020804.4 | NP_065855.1 | 8 | 1224 | ||
XM_011514541.1 | XP_011512843.1 | 8 | 1079 | ||
GenBank mRNA | AB037800.2 | - | 8 | 1181 | |
AF242529.1 | - | 8 | 1147 | ||
AK289956.1 | - | 8 | 1209 | ||
AK294132.1 | - | 8 | 1211 | ||
AL834211.1 | - | 8 | 1157 | ||
BC040228.1 | - | 8 | 1196 | 5173129 | |
BC128219.1 | - | 5 | 604 |