Gene Transcripts
- Gene Symbol
- ZDHHC9
- Entrez Gene ID
- 51114
- Gene Name
- zinc finger DHHC-type containing 9
- Gene Aliases
- CGI89, CXorf11, DHHC9, MMSA1, MRXSZ, ZDHHC10, ZNF379, ZNF380
- Chromosome Location
- Chr. X: 129803288 - 129843934 on Build GRCh38
- UniGene ID
- Hs.193566
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001008222.2 |
NP_001008223.1
|
6, 7 |
1142 |
|
NM_016032.3 |
NP_057116.2
|
7, 8 |
1122 |
|
GenBank mRNA |
AB209807.1 |
- |
6, 7 |
1061 |
|
AF151847.1 |
- |
7, 8 |
1010 |
|
AK001112.1 |
- |
7, 8 |
1072 |
|
AK001424.1 |
- |
4, 5 |
456 |
|
AK001524.1 |
- |
6, 7 |
873 |
|
AK096967.1 |
- |
7, 8 |
773 |
|
AL161962.1 |
- |
7, 8 |
999 |
|
AY358558.1 |
- |
7, 8 |
1054 |
|
AY952881.8 |
- |
7, 8 |
1094 |
|
BC000035.2 |
- |
6, 7 |
1016 |
3506601 |
BC003128.1 |
- |
7, 8 |
1060 |
2957911 |
BC006200.2 |
- |
7, 8 |
1056 |
2964425 |
BC012826.2 |
- |
7, 8 |
1056 |
2964425 |
View Full Product Details