Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001135186.1 | NP_001128658.1 | 3 | 912 | |
NM_016428.2 | NP_057512.1 | 3 | 930 | ||
XM_005257429.2 | XP_005257486.1 | 3 | 575 | ||
XM_011524873.1 | XP_011523175.1 | 3 | 575 | ||
XM_017024721.1 | XP_016880210.1 | 3 | 575 | ||
GenBank mRNA | AB037886.1 | - | 3 | 930 | |
AL136709.1 | - | 3 | 731 | ||
BC007780.2 | - | 3 | 436 | 4309524 | |
BI768790.1 | - | 3 | 419 |