Gene Transcripts

Gene Symbol
AIG1
Entrez Gene ID
51390
Gene Name
androgen induced 1
Gene Aliases
AIG-1, dJ95L4.1
Chromosome Location
Chr. 6: 143059363 - 143343594 on Build GRCh38
UniGene ID
Hs.567501
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286587.1 NP_001273516.1 2 71
NM_001286588.1 NP_001273517.1 1 128
NM_001286589.1 NP_001273518.1 1 128
NM_016108.3 NP_057192.2 1 128
XM_011535881.2 XP_011534183.1 1 110
XM_011535882.2 XP_011534184.1 1 109
XM_011535883.2 XP_011534185.1 1 109
XM_011535884.2 XP_011534186.1 1 111
XM_011535885.2 XP_011534187.1 1 109
XM_017010927.1 XP_016866416.1 1 127
XM_017010928.1 XP_016866417.1 1 109
GenBank mRNA AF151861.1 - 2 74
AK001347.1 - 1 69
AK222475.1 - 1 90
BC025278.1 - 1 59 4859216
BX640703.1 - 1 70

View Full Product Details