Gene Transcripts
- Gene Symbol
- ZCCHC17
- Entrez Gene ID
- 51538
- Gene Name
- zinc finger CCHC-type containing 17
- Gene Aliases
- HSPC251, PS1D, pNO40
- Chromosome Location
- Chr. 1: 31296982 - 31365764 on Build GRCh38
- UniGene ID
- Hs.524094
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001282566.1 |
NP_001269495.1
|
2, 3 |
296 |
|
NM_001282567.1 |
NP_001269496.1
|
2, 3 |
158 |
|
NM_001282568.1 |
NP_001269497.1
|
3, 4 |
417 |
|
NM_001282569.1 |
NP_001269498.1
|
4, 5 |
349 |
|
NM_001282570.1 |
NP_001269499.1
|
2, 3 |
158 |
|
NM_001282572.1 |
NP_001269501.1
|
3, 4 |
279 |
|
NM_001282573.1 |
NP_001269502.1
|
4, 5 |
349 |
|
NM_001282574.1 |
NP_001269503.1
|
3, 4 |
279 |
|
NM_016505.3 |
NP_057589.2
|
3, 4 |
279 |
|
XM_006710681.3 |
XP_006710744.1
|
4, 5 |
423 |
|
XM_011541567.2 |
XP_011539869.1
|
3, 4 |
295 |
|
GenBank mRNA |
AF151077.1 |
- |
3, 4 |
208 |
|
AF151085.1 |
- |
3, 4 |
237 |
|
AF247661.2 |
- |
2, 3 |
148 |
|
AK001929.1 |
- |
3, 4 |
229 |
|
AK022506.1 |
- |
3, 4 |
266 |
|
AK024049.1 |
- |
3, 4 |
255 |
|
AK302248.1 |
- |
4, 5 |
299 |
|
AK302343.1 |
- |
2, 3 |
104 |
|
AK307478.1 |
- |
2, 3 |
135 |
|
BC000685.2 |
- |
3, 4 |
317 |
|
BC007446.2 |
- |
3, 4 |
215 |
3009823 |
BC050609.1 |
- |
3, 4 |
219 |
6527025 |
BF309574.1 |
- |
3, 4 |
230 |
|
CN423139.1 |
- |
3, 4 |
216 |
|
DA737244.1 |
- |
4, 5 |
299 |
|
DC366453.1 |
- |
3, 4 |
417 |
|
View Full Product Details