Gene Transcripts

Gene Symbol
NMRK1
Entrez Gene ID
54981
Gene Name
nicotinamide riboside kinase 1
Gene Aliases
C9orf95, NRK1, bA235O14.2
Chromosome Location
Chr. 9: 75060577 - 75088319 on Build GRCh38
UniGene ID
Hs.494186
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127603.1 NP_001121075.1 3 324
NM_017881.2 NP_060351.1 3 324
NR_023352.1 4 581
XM_006717162.1 XP_006717225.1 4 609
XM_006717163.2 XP_006717226.1 4 644
XM_011518817.1 XP_011517119.1 3 434
XM_011518818.1 XP_011517120.1 3 956
XM_011518819.2 XP_011517121.1 3 956
XM_017014868.1 XP_016870357.1 5 930
XM_017014869.1 XP_016870358.1 4 1237
XM_017014870.1 XP_016870359.1 3 980
XM_017014871.1 XP_016870360.1 4 609
XM_017014872.1 XP_016870361.1 5 930
XM_017014873.1 XP_016870362.1 5 901
XM_017014874.1 XP_016870363.1 4 644
GenBank mRNA AK000566.1 - 3 289
AK023413.1 - 3 316
AK097144.1 - 4 554
AY611480.1 - 2 79
BC001366.2 - 3 232 3051165
BC036804.1 - 3 324 5727308
CB135264.1 - 3 324

View Full Product Details