Gene Transcripts

Gene Symbol
TMEM19
Entrez Gene ID
55266
Gene Name
transmembrane protein 19
Gene Aliases
Chromosome Location
Chr. 12: 71686098 - 71704060 on Build GRCh38
UniGene ID
Hs.644802
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018279.3 NP_060749.2 1, 2 715
GenBank mRNA AK001798.1 - 1, 2 317
AK223148.1 - 1, 2 313
AK315587.1 - 1, 2 274
BC008596.1 - 1, 2 715 4154303

View Full Product Details