Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018229.3 | NP_060699.3 | 3 | 1344 | |
NR_026895.1 | 2 | 698 | |||
XM_006720196.2 | XP_006720259.1 | 3 | 973 | ||
XM_011536940.2 | XP_011535242.1 | 4 | 1316 | ||
GenBank mRNA | AF094583.1 | - | 2 | 309 | |
AK001675.1 | - | 3 | 1189 | ||
AK172824.1 | - | 2 | 406 | ||
AK225345.1 | - | 3 | 1191 | ||
AK303999.1 | - | 4 | 1218 | ||
AL136685.1 | - | 3 | 1323 |