Gene Transcripts

Gene Symbol
EMSY
Entrez Gene ID
56946
Gene Name
EMSY, BRCA2 interacting transcriptional repressor
Gene Aliases
C11orf30, GL002
Chromosome Location
Chr. 11: 76444928 - 76552901 on Build GRCh38
UniGene ID
Hs.352588
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300942.1 NP_001287871.1 12, 13 1866
NM_001300943.1 NP_001287872.1 11, 12 1824
NM_001300944.1 NP_001287873.1 12, 13 1866
NM_020193.4 NP_064578.2 11, 12 1821
XM_005274112.4 XP_005274169.1 11, 12 1810
XM_006718637.3 XP_006718700.1 12, 13 1963
XM_006718638.3 XP_006718701.1 11, 12 1834
XM_006718639.3 XP_006718702.1 10, 11 1681
XM_011545173.2 XP_011543475.1 12, 13 1963
XM_011545174.2 XP_011543476.1 12, 13 1960
XM_011545175.2 XP_011543477.1 11, 12 1834
XM_011545177.2 XP_011543479.1 11, 12 1810
XM_011545178.2 XP_011543480.1 12, 13 1963
XM_011545179.2 XP_011543481.1 10, 11 1681
XM_017018021.1 XP_016873510.1 12, 13 1963
XM_017018022.1 XP_016873511.1 12, 13 1809
GenBank mRNA AJ430203.1 - 10, 11 1678
AK125114.1 - 10, 11 1535
BC117265.1 - 10, 11 1720 40125816
BC143370.1 - 11, 12 1759 9051875
BC143374.1 - 11, 12 1762 9051879
BC143376.1 - 11, 12 1762 9051881

View Full Product Details