Gene Transcripts

Gene Symbol
BBX
Entrez Gene ID
56987
Gene Name
BBX, HMG-box containing
Gene Aliases
ARTC1, HBP2, HSPC339, MDS001
Chromosome Location
Chr. 3: 107522936 - 107811329 on Build GRCh38
UniGene ID
Hs.124366
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142568.2 NP_001136040.1 11 1305
NM_020235.6 NP_064620.2 11 1305
XM_005247642.3 XP_005247699.1 11 1221
XM_005247643.3 XP_005247700.1 11 1233
XM_005247644.3 XP_005247701.1 12 1396
XM_011512999.2 XP_011511301.1 11 1305
XM_011513000.1 XP_011511302.1 11 1433
XM_011513001.1 XP_011511303.1 11 1273
XM_011513002.2 XP_011511304.1 11 1305
XM_011513003.2 XP_011511305.1 11 1305
XM_011513004.2 XP_011511306.1 11 1433
XM_011513005.2 XP_011511307.1 11 1305
XM_017006881.1 XP_016862370.1 13 1446
XM_017006882.1 XP_016862371.1 12 1396
GenBank mRNA AF161457.1 - 11 1285
AF276948.1 - 8 1003
AF276949.1 - 11 1236
AF283775.2 - 11 1299
AF454941.1 - 11 1298
AF454942.1 - 9 1178
AK057010.1 - 11 1251
AK291757.1 - 11 1251
AK307668.1 - 10 1345
AK314434.1 - 11 1256
AL136769.1 - 9 1178
BC110903.1 - 9 1215

View Full Product Details