Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020974.2 | NP_066025.2 | 19 | 2441 | |
XM_005253032.4 | XP_005253089.2 | 20 | 2608 | ||
XM_005253033.4 | XP_005253090.2 | 19 | 2521 | ||
XM_005253034.4 | XP_005253091.2 | 19 | 2485 | ||
XM_005253035.4 | XP_005253092.2 | 20 | 2608 | ||
XM_005253036.4 | XP_005253093.2 | 17 | 2281 | ||
XM_005253037.4 | XP_005253094.2 | 16 | 2143 | ||
XM_011520247.2 | XP_011518549.1 | 18 | 2404 | ||
XM_017018080.1 | XP_016873569.1 | 18 | 2398 | ||
XM_017018082.1 | XP_016873571.1 | 17 | 2281 | ||
GenBank mRNA | AK092553.1 | - | 7 | 972 | |
AK123039.1 | - | 19 | 2437 |