Gene Transcripts

Gene Symbol
INIP
Entrez Gene ID
58493
Gene Name
INTS3 and NABP interacting protein
Gene Aliases
C9orf80, HSPC043, MISE, SOSSC, SSBIP1, hSSBIP1
Chromosome Location
Chr. 9: 112686506 - 112718107 on Build GRCh38
UniGene ID
Hs.658575
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021218.1 NP_067041.1 2, 3 191
GenBank mRNA AF161409.1 - 2, 3 159
AF161411.2 - 2, 3 124
AK000637.1 - 2, 3 194
AK310007.1 - 2, 3 191
BC013097.1 - 50
BC014881.1 - 2, 3 191 3907074
BC065210.1 - 2, 3 189 3456693

View Full Product Details