Gene Transcripts

Gene Symbol
RRAGC
Entrez Gene ID
64121
Gene Name
Ras related GTP binding C
Gene Aliases
GTR2, RAGC, TIB929
Chromosome Location
Chr. 1: 38838197 - 38859823 on Build GRCh38
UniGene ID
Hs.532461
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271851.1 NP_001258780.1 3 664
NM_022157.3 NP_071440.1 3 766
GenBank mRNA AF272035.1 - 3 594
AF323609.1 - 3 589
AK298573.1 - 2 289
AK298788.1 - 3 560
AK315791.1 - 3 691
BC016668.1 - 3 611 4342448

View Full Product Details