Gene Transcripts
- Gene Symbol
- OSGEPL1
- Entrez Gene ID
- 64172
- Gene Name
- O-sialoglycoprotein endopeptidase-like 1
- Gene Aliases
- Qri7
- Chromosome Location
- Chr. 2: 189746660 - 189763198 on Build GRCh38
- UniGene ID
- Hs.60772
- Species
- Homo sapiens
- Gene Symbol
- ANKAR
- Entrez Gene ID
- 150709
- Gene Name
- ankyrin and armadillo repeat containing
- Gene Aliases
- Chromosome Location
- Chr. 2: 189674292 - 189763059 on Build GRCh38
- UniGene ID
- Hs.60772
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK027836.1 |
- |
1 |
1098 |
|
View Full Product Details