Gene Transcripts
- Gene Symbol
- RMND5B
- Entrez Gene ID
- 64777
- Gene Name
- required for meiotic nuclear division 5 homolog B
- Gene Aliases
- GID2, GID2B
- Chromosome Location
- Chr. 5: 178130532 - 178148570 on Build GRCh38
- UniGene ID
- Hs.27222
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001288794.1 |
NP_001275723.1
|
10, 11 |
1347 |
|
NM_001288795.1 |
NP_001275724.1
|
9, 10 |
1196 |
|
NM_022762.4 |
NP_073599.2
|
9, 10 |
1207 |
|
XM_005265969.3 |
XP_005266026.1
|
8, 9 |
1027 |
|
XM_005265971.4 |
XP_005266028.1
|
9, 10 |
1774 |
|
XM_005265972.3 |
XP_005266029.1
|
8, 9 |
1154 |
|
XM_017009727.1 |
XP_016865216.1
|
8, 9 |
1818 |
|
XM_017009728.1 |
XP_016865217.1
|
8, 9 |
1485 |
|
GenBank mRNA |
AK025971.1 |
- |
6, 7 |
752 |
|
AK093481.1 |
- |
9, 10 |
1148 |
|
AK093622.1 |
- |
1, 2 |
1664 |
|
AK094065.1 |
- |
6, 7 |
1354 |
|
AK122692.1 |
- |
4, 5 |
2070 |
|
AK125337.1 |
- |
10, 11 |
1294 |
|
AL137381.1 |
- |
3, 4 |
320 |
|
AL713670.1 |
- |
8, 9 |
1070 |
|
AY359092.1 |
- |
6, 7 |
2065 |
|
BC009911.2 |
- |
9, 10 |
1141 |
2820100 |
DQ494789.1 |
- |
9, 10 |
1141 |
|
View Full Product Details