Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023080.2 | NP_075568.1 | 2 | 247 | |
GenBank mRNA | AK024642.1 | - | 2 | 245 | |
BC010001.2 | - | 2 | 207 | 4135341 | |
BX648996.1 | - | 1 | 307 | ||
CR457330.1 | - | 1 | 193 | ||
DA467518.1 | - | 2 | 247 |