Gene Transcripts
- Gene Symbol
- SPATA5L1
- Entrez Gene ID
- 79029
- Gene Name
- spermatogenesis associated 5 like 1
- Gene Aliases
- Chromosome Location
- Chr. 15: 45400202 - 45421419 on Build GRCh38
- UniGene ID
- Hs.369657
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024063.2 |
NP_076968.2
|
6, 7 |
2076 |
|
NR_027635.1 |
|
6, 7 |
2106 |
|
NR_136645.1 |
|
6, 7 |
2083 |
|
NR_136646.1 |
|
7, 8 |
2301 |
|
NR_136647.1 |
|
7, 8 |
2268 |
|
NR_136648.1 |
|
7, 8 |
2271 |
|
GenBank mRNA |
AF086107.1 |
- |
1, 2 |
117 |
|
AK023232.1 |
- |
6, 7 |
2052 |
|
AK225463.1 |
- |
6, 7 |
2022 |
|
BC000981.2 |
- |
6, 7 |
2009 |
2985725 |
BC051861.2 |
- |
6, 7 |
2021 |
|
View Full Product Details