Gene Transcripts
- Gene Symbol
- RNF219
- Entrez Gene ID
- 79596
- Gene Name
- ring finger protein 219
- Gene Aliases
- C13orf7
- Chromosome Location
- Chr. 13: 78614286 - 78659179 on Build GRCh38
- UniGene ID
- Hs.567576
- Species
- Homo sapiens
- Gene Symbol
- RNF219-AS1
- Entrez Gene ID
- 100874222
- Gene Name
- RNF219 antisense RNA 1
- Gene Aliases
- POU4F1-AS1
- Chromosome Location
- Chr. 13: 78054855 - 78617325 on Build GRCh38
- UniGene ID
- Hs.493062
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_047001.1 |
|
6 |
1070 |
|
GenBank mRNA |
AK095779.1 |
- |
6 |
1071 |
|
View Full Product Details