Gene Transcripts

Gene Symbol
RNF219
Entrez Gene ID
79596
Gene Name
ring finger protein 219
Gene Aliases
C13orf7
Chromosome Location
Chr. 13: 78614286 - 78659179 on Build GRCh38
UniGene ID
Hs.567576
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024546.3 NP_078822.3 4, 5 603
XM_011535225.1 XP_011533527.1 3, 4 453
XM_017020743.1 XP_016876232.1 4, 5 2398
GenBank mRNA AK098640.1 - 4, 5 615
AL136651.1 - 4, 5 575
BC136764.1 - 4, 5 598 9020754
BC136765.1 - 4, 5 598 9020755
BI550796.1 - 4, 5 521
CR936752.1 - 4, 5 580
CR936754.1 - 4, 5 590
CR936764.1 - 4, 5 577

View Full Product Details