Gene Transcripts

Gene Symbol
C7orf69
Entrez Gene ID
80099
Gene Name
chromosome 7 open reading frame 69
Gene Aliases
Chromosome Location
Chr. 7: 47795291 - 47819847 on Build GRCh38
UniGene ID
Hs.287647
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001302627.1 NP_001289556.1 4 503
NM_025031.2 NP_079307.2 3 359
GenBank mRNA AK024728.1 - 3 359
BC113679.1 - 3 350 8322731
BC113681.1 - 3 350 8322733
CA420605.1 - 1 317
DA711092.1 - 4 466

View Full Product Details