Gene Transcripts

Gene Symbol
BBOF1
Entrez Gene ID
80127
Gene Name
basal body orientation factor 1
Gene Aliases
C14orf45, CCDC176
Chromosome Location
Chr. 14: 74019350 - 74078351 on Build GRCh38
UniGene ID
Hs.644621
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025057.2 NP_079333.2 10, 11 1596
XM_005268092.3 XP_005268149.1 10, 11 1602
XM_005268097.3 XP_005268154.1 8, 9 963
XM_005268098.4 XP_005268155.1 8, 9 1235
XM_011537170.2 XP_011535472.1 11, 12 1710
XM_011537171.2 XP_011535473.1 11, 12 1710
XM_011537172.1 XP_011535474.1 11, 12 1710
XM_011537174.2 XP_011535476.1 11, 12 1710
XM_011537175.2 XP_011535477.1 10, 11 1566
XM_011537176.2 XP_011535478.1 12, 13 1694
XM_011537177.2 XP_011535479.1 10, 11 1414
XM_011537178.2 XP_011535480.1 10, 11 1385
XM_011537179.2 XP_011535481.1 9, 10 1270
XM_017021659.1 XP_016877148.1 10, 11 1602
XM_017021660.1 XP_016877149.1 9, 10 1318
XM_017021661.1 XP_016877150.1 9, 10 1370
XM_017021662.1 XP_016877151.1 8, 9 1154
XM_017021663.1 XP_016877152.1 8, 9 963
GenBank mRNA AK026842.1 - 3, 4 629
AK097531.1 - 8, 9 1274
AL834487.1 - 5, 6 871
BC104979.1 - 3, 4 627
BC104981.1 - 3, 4 627

View Full Product Details