Gene Transcripts

Gene Symbol
OR2C3
Entrez Gene ID
81472
Gene Name
olfactory receptor family 2 subfamily C member 3
Gene Aliases
OR2C4, OR2C5P, OST742
Chromosome Location
Chr. 1: 247530132 - 247533839 on Build GRCh38
UniGene ID
Hs.730218
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198074.4 NP_932340.3 2 386
GenBank mRNA BC030717.1 - 2 384

View Full Product Details