Gene Transcripts

Gene Symbol
THAP2
Entrez Gene ID
83591
Gene Name
THAP domain containing 2
Gene Aliases
Chromosome Location
Chr. 12: 71663897 - 71680648 on Build GRCh38
UniGene ID
Hs.245798
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031435.3 NP_113623.1 3 977
GenBank mRNA AK313452.1 - 3 539
AL136607.1 - 3 874
BC008358.1 - 3 717 3606376
CR533485.1 - 3 364

View Full Product Details