Gene Transcripts

Gene Symbol
C10orf11
Entrez Gene ID
83938
Gene Name
chromosome 10 open reading frame 11
Gene Aliases
CDA017
Chromosome Location
Chr. 10: 75431646 - 76559220 on Build GRCh38
UniGene ID
Hs.118161
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032024.4 NP_114413.1 1 243
GenBank mRNA AF267860.1 - 1 244

View Full Product Details