Gene Transcripts
- Gene Symbol
- ARRDC1
- Entrez Gene ID
- 92714
- Gene Name
- arrestin domain containing 1
- Gene Aliases
- Chromosome Location
- Chr. 9: 137605640 - 137615360 on Build GRCh38
- UniGene ID
- Hs.12999
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB209523.1 |
- |
3 |
2674 |
|
- Gene Symbol
- ARRDC1-AS1
- Entrez Gene ID
- 85026
- Gene Name
- ARRDC1 antisense RNA 1
- Gene Aliases
- C9orf37
- Chromosome Location
- Chr. 9: 137615332 - 137618895 on Build GRCh38
- UniGene ID
- Hs.522520
- Species
- Homo sapiens
View Full Product Details