Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033087.3 | NP_149078.1 | 2 | 1295 | |
NR_024532.1 | 3 | 1454 | |||
GenBank mRNA | AB161356.1 | - | 2 | 1252 | |
AK027417.1 | - | 2 | 1247 | ||
AK027814.1 | - | 1 | 2047 | ||
AK074704.1 | - | 2 | 1247 | ||
AK074988.1 | - | 3 | 1432 | ||
AK075172.1 | - | 2 | 1271 | ||
AK075189.1 | - | 1 | 515 | ||
AY358697.1 | - | 2 | 1039 | ||
BC017876.1 | - | 2 | 1245 | 4698763 |