Gene Transcripts

Gene Symbol
SPSB3
Entrez Gene ID
90864
Gene Name
splA/ryanodine receptor domain and SOCS box containing 3
Gene Aliases
C16orf31, SSB3
Chromosome Location
Chr. 16: 1776712 - 1782605 on Build GRCh38
UniGene ID
Hs.592080
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001324081.1 NP_001311010.1 4 435
NM_080861.3 NP_543137.2 4 457
XM_005255673.4 XP_005255730.1 4 647
GenBank mRNA AF403028.1 - 3 366
AK057850.1 - 419
AK074190.1 - 4 421
BC007588.1 - 4 403 3342825
BC024244.1 - 4 421 4798873
BC035065.1 - 4 414
BC041897.1 - 3 1231 5298729
BC047441.1 - 4 399 5303706
BC065515.1 - 3 542
BI768095.1 - 4 411
BM836743.1 - 4 482

Gene Symbol
EME2
Entrez Gene ID
197342
Gene Name
essential meiotic structure-specific endonuclease subunit 2
Gene Aliases
SLX2B, gs125
Chromosome Location
Chr. 16: 1773222 - 1776714 on Build GRCh38
UniGene ID
Hs.7247
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK074080.2 - 7 3245

View Full Product Details