Gene Transcripts

Gene Symbol
STK11IP
Entrez Gene ID
114790
Gene Name
serine/threonine kinase 11 interacting protein
Gene Aliases
LIP1, LKB1IP, STK11IP1
Chromosome Location
Chr. 2: 219597851 - 219616451 on Build GRCh38
UniGene ID
Hs.22410
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052902.3 NP_443134.3 8 742
XM_011510526.1 XP_011508828.1 8 722
XM_011510527.2 XP_011508829.1 8 722
XM_011510529.1 XP_011508831.1 8 722
GenBank mRNA AB067485.1 - 6 420
AF450267.1 - 8 696
AK091963.1 - 7 624
AK295609.1 - 8 725
AK296632.1 - 8 722
AK300611.1 - 8 725
BC034051.1 - 8 719 4819148

View Full Product Details