Gene Transcripts
- Gene Symbol
- CCDC43
- Entrez Gene ID
- 124808
- Gene Name
- coiled-coil domain containing 43
- Gene Aliases
- Chromosome Location
- Chr. 17: 44677437 - 44689797 on Build GRCh38
- UniGene ID
- Hs.579115
- Species
- Homo sapiens
- Gene Symbol
- MEIOC
- Entrez Gene ID
- 284071
- Gene Name
- meiosis specific with coiled-coil domain
- Gene Aliases
- C17orf104
- Chromosome Location
- Chr. 17: 44656394 - 44677086 on Build GRCh38
- UniGene ID
- Hs.349758
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK097747.1 |
- |
10 |
1597 |
|
View Full Product Details