Gene Transcripts

Gene Symbol
EFCAB13
Entrez Gene ID
124989
Gene Name
EF-hand calcium binding domain 13
Gene Aliases
C17orf57
Chromosome Location
Chr. 17: 47323961 - 47441312 on Build GRCh38
UniGene ID
Hs.463303
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152347.4 NP_689560.3 9, 10 1102
GenBank mRNA AK097661.1 - 6, 7 661
BC036407.2 - 9, 10 1082 5266195

View Full Product Details