Gene Transcripts
- Gene Symbol
- OCIAD2
- Entrez Gene ID
- 132299
- Gene Name
- OCIA domain containing 2
- Gene Aliases
- Chromosome Location
- Chr. 4: 48885380 - 48906828 on Build GRCh38
- UniGene ID
- Hs.95835
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001014446.2 |
NP_001014446.1
|
4, 5 |
441 |
|
NM_001286773.1 |
NP_001273702.1
|
4, 5 |
464 |
|
NM_001286774.1 |
NP_001273703.1
|
3, 4 |
344 |
|
NM_152398.3 |
NP_689611.1
|
4, 5 |
441 |
|
GenBank mRNA |
AK298305.1 |
- |
4, 5 |
410 |
|
AY472117.1 |
- |
4, 5 |
443 |
|
BC032808.1 |
- |
4, 5 |
412 |
4454708 |
BI546814.1 |
- |
4, 5 |
446 |
|
BM782144.1 |
- |
4, 5 |
411 |
|
BU508058.1 |
- |
3, 4 |
232 |
|
CR749310.1 |
- |
4, 5 |
386 |
|
W68799.1 |
- |
4, 5 |
393 |
|
View Full Product Details