Gene Transcripts

Gene Symbol
C4orf33
Entrez Gene ID
132321
Gene Name
chromosome 4 open reading frame 33
Gene Aliases
Chromosome Location
Chr. 4: 129093674 - 129112688 on Build GRCh38
UniGene ID
Hs.567679
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099783.1 NP_001093253.1 2 139
NM_173487.2 NP_775758.2 2 411
XM_005262734.4 XP_005262791.1 2 406
XM_017007722.1 XP_016863211.1 2 1468
XM_017007723.1 XP_016863212.1 2 1218
XM_017007724.1 XP_016863213.1 2 394
GenBank mRNA AK091022.1 - 2 400
AK309229.1 - 2 385
BC032582.1 - 2 342 5494700
BE566349.1 - 2 94
BP871612.1 - 2 411
BQ549846.1 - 2 170
BX538164.1 - 2 352
CN483524.1 - 2 139

View Full Product Details